missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoisoquinolin-3-amine, 97%, Thermo Scientific™

Catalog Number 10502462
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
13130-79-5
C9H7BrN2
223.073
MFCD00102193
PSQUIUNIVDKHJK-UHFFFAOYSA-N
3-amino-1-bromoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromoisoquinolin-3-ylamine, 1-bromo-3-isoquinolylamine, 3-amino-1-bromo-isoquinoline, 1-bromoisoquinoline-3-amine, acmc-1by0f, 1-bromo-3-aminoisoquinoline, 1-bromanylisoquinolin-3-amine, 3-isoquinolinamine, 1-bromo
289845
1-bromoisoquinolin-3-amine
C1=CC=C2C(=C1)C=C(N=C2Br)N
This item is not returnable. View return policy
13130-79-5
C9H7BrN2
223.073
MFCD00102193
PSQUIUNIVDKHJK-UHFFFAOYSA-N
3-amino-1-bromoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromoisoquinolin-3-ylamine, 1-bromo-3-isoquinolylamine, 3-amino-1-bromo-isoquinoline, 1-bromoisoquinoline-3-amine, acmc-1by0f, 1-bromo-3-aminoisoquinoline, 1-bromanylisoquinolin-3-amine, 3-isoquinolinamine, 1-bromo
289845
1-bromoisoquinolin-3-amine
C1=CC=C2C(=C1)C=C(N=C2Br)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 1-Bromoisoquinolin-3-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'