missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane, 98%, Thermo Scientific Chemicals

Catalog Number p-4229108
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
143-15-7
C12H25Br
249.23
PBLNBZIONSLZBU-UHFFFAOYSA-N
dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
8919
1-bromododecane
CCCCCCCCCCCCBr
This item is not returnable. View return policy
143-15-7
C12H25Br
249.23
PBLNBZIONSLZBU-UHFFFAOYSA-N
dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
8919
1-bromododecane
CCCCCCCCCCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -11°C to 9°C
Density 1.0300g/mL
Boiling Point 134.0°C to 135.0°C (6.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11Br
Refractive Index 1.4570 to 1.4590
Quantity 250 mL
Beilstein 01,II,133
Merck Index 15,5444
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble
Formula Weight 249.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromododecane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'