missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane, 98%, ACROS Organics™

Brand:  Acros Organics 106910025

Code : 29

Additional Details : CAS Number : 143-15-7 Weight : 2.62500kg

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Product Code. 11994481

  • £130.90 / 2.50 L


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

1-Bromododecane
97.5
98%
C12H25Br
249.23
143-15-7
100.0
98%
CH3(CH2)11Br
Lauryl bromide

Specifications

1-Bromododecane
97.5
Authentic
1.4570 to 1.4590
1.0300g/mL
− 11.0° to 9.0°C
98%
98%
110°C
CH3(CH2)11Br
Lauryl bromide
143-15-7
100.0
Glass bottle
0.1% max. (K.F.)
Clear
Liquid
134.0° to 135.0°C (6.0 mmHg)
2.5L
C12H25Br
249.23
Videos
SDS