missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-5-fluoropentane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as ?-opioid receptor imaging agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.
Solubility
Soluble in methanol (in methanol). Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.
Solubility
Soluble in methanol (in methanol). Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | Colorless |
Density | 1.360 g/mL |
Boiling Point | 162°C |
Linear Formula | F(CH2)5Br |
Refractive Index | 1.4406 |
Quantity | 250 mg |
Beilstein | 1697258 |
Solubility Information | Soluble in methanol (in methanol). Slightly soluble in water. |
Formula Weight | 169.04 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'