missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluorobutane, 97%, Thermo Scientific Chemicals

Catalog Number 15431469
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
462-72-6
C4H8BrF
155.01
MFCD00209551
WMCUHRDQSHQNRW-UHFFFAOYSA-N
10022
1-bromo-4-fluorobutane
C(CCBr)CF
This item is not returnable. View return policy
462-72-6
C4H8BrF
155.01
MFCD00209551
WMCUHRDQSHQNRW-UHFFFAOYSA-N
10022
1-bromo-4-fluorobutane
C(CCBr)CF

1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

Solubility
Sparingly soluble in water at 25°C 0.48 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.444 g/mL
Boiling Point 134°C to 135°C
Flash Point 45°C (113°F)
Linear Formula FCH2CH2CH2CH2Br
Refractive Index 1.439
Quantity 5 g
UN Number UN1993
Beilstein 1731414
Synonym 4-fluorobutyl bromide, butane, 1-bromo-4-fluoro, butane,1-bromo-4-fluoro, fluorobutyl bromide, acmc-20aoww, 4-bromo-1-fluorobutane, 1-bromo-4-fluoro-butane, 4-01-00-00263 beilstein handbook reference, 1-bromo-4-fluorobutane, 1-bromo-4-fluoro-butan;4-fluoro-1-bromobutane
Solubility Information Sparingly soluble in water at 25°C 0.48g/L.
Formula Weight 155.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-fluorobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'