missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluorobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-7081595
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
460-00-4
C6H4BrF
175.00
MFCD00000342
AITNMTXHTIIIBB-UHFFFAOYSA-N
4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
9993
1-bromo-4-fluorobenzene
FC1=CC=C(Br)C=C1
This item is not returnable. View return policy
460-00-4
C6H4BrF
175.00
MFCD00000342
AITNMTXHTIIIBB-UHFFFAOYSA-N
4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
9993
1-bromo-4-fluorobenzene
FC1=CC=C(Br)C=C1

1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

Solubility
Soluble in water(0.14g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point -16°C
Density 1.604
Boiling Point 151°C to 153°C
Flash Point 60°C (140°F)
Odor Aromatic
Refractive Index 1.527
Quantity 100 g
UN Number UN1993
Beilstein 774102
Solubility Information Soluble in water(0.14g/L).
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-fluorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'