missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(difluoromethyl)benzene, 97%, Thermo Scientific™

Catalog Number p-4228433
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
51776-71-7
C7H5BrF2
207.02
MFCD06657964
HUSPSWKWFREKSS-UHFFFAOYSA-N
1-bromo-4-difluoromethyl benzene, 1-bromo-4-difluoromethyl-benzene, benzene, 1-bromo-4-difluoromethyl, 4-bromo-alpha,alpha-difluorotoluene, 4-bromobenzal fluoride, pubchem23296, acmc-1aukw, 1-bromo-4-difluoro-methyl-benzene
2761183
1-bromo-4-(difluoromethyl)benzene
FC(F)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
51776-71-7
C7H5BrF2
207.02
MFCD06657964
HUSPSWKWFREKSS-UHFFFAOYSA-N
1-bromo-4-difluoromethyl benzene, 1-bromo-4-difluoromethyl-benzene, benzene, 1-bromo-4-difluoromethyl, 4-bromo-alpha,alpha-difluorotoluene, 4-bromobenzal fluoride, pubchem23296, acmc-1aukw, 1-bromo-4-difluoro-methyl-benzene
2761183
1-bromo-4-(difluoromethyl)benzene
FC(F)C1=CC=C(Br)C=C1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 74.0°C to 76.0°C (12.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-(difluoromethyl)benzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-4-(difluoromethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'