missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane, 99%, Thermo Scientific Chemicals

Catalog Number p-7081424
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
6940-78-9
C4H8BrCl
171.462
MFCD00001010
NIDSRGCVYOEDFW-UHFFFAOYSA-N
butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
81364
1-bromo-4-chlorobutane
C(CCBr)CCl
This item is not returnable. View return policy
6940-78-9
C4H8BrCl
171.462
MFCD00001010
NIDSRGCVYOEDFW-UHFFFAOYSA-N
butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
81364
1-bromo-4-chlorobutane
C(CCBr)CCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.489
Boiling Point 175°C to 176°C
Flash Point 60°C (140°F)
Odor Aromatic
Refractive Index 1.4875
Quantity 25 g
UN Number UN1993
Beilstein 1098275
Formula Weight 171.47
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-chlorobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'