missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081953
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy
106-39-8
C6H4BrCl
191.45
MFCD00000600
NHDODQWIKUYWMW-UHFFFAOYSA-N
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
7806
1-bromo-4-chlorobenzene
ClC1=CC=C(Br)C=C1
This item is not returnable. View return policy
106-39-8
C6H4BrCl
191.45
MFCD00000600
NHDODQWIKUYWMW-UHFFFAOYSA-N
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
7806
1-bromo-4-chlorobenzene
ClC1=CC=C(Br)C=C1

1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 0.045g/L.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 63°C to 67°C
Density 1.651
Boiling Point 196°C
Quantity 100 g
Odor Aromatic
UN Number UN2811
Beilstein 1099014
Solubility Information Soluble in water 0.045g/L.
Formula Weight 191.46
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-chlorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'