missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chloro-2-fluorobenzene, 97%, ACROS Organics™

Brand:  Acros Organics 294050050

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1996-29-8 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10672593

  • £24.90 / 5g


EU Stock 2
Add to basket

Specifications Description & Specifications

Chemical Identifiers

1-Bromo-4-chloro-2-fluorobenzene
1996-29-8
100.0
97%
MFCD00079708
97%
96.0
≥96.0%
209.44

Specifications

1-Bromo-4-chloro-2-fluorobenzene
1996-29-8
96.0
Glass bottle
Clear colorless to pale yellow
Liquid
97%
MFCD00079708
97%
Authentic
100.0
1.5540 to 1.5560 (20°C, 589nm)
5g
≥96.0%
209.44
Documentation

Technical Documents