missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chloro-2-fluorobenzene, 97%, Thermo Scientific™

Catalog Number p-2689521
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1996-29-8
C6H3BrClF
209.44
MFCD00079708
FPNVMCMDWZNTEU-UHFFFAOYSA-N
4-chloro-2-fluorobromobenzene, 2-fluoro-4-chlorobromobenzene, 2-bromo-5-chlorofluorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 1-bromo-2-fluoro-4-chlorobenzene, 2-bromo-5-chloro-1-fluorobenzene, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209, acmc-1bovk, 2-bromo-5-chlorofluorbenzene
137275
1-bromo-4-chloro-2-fluorobenzene
FC1=CC(Cl)=CC=C1Br
This item is not returnable. View return policy
1996-29-8
C6H3BrClF
209.44
MFCD00079708
FPNVMCMDWZNTEU-UHFFFAOYSA-N
4-chloro-2-fluorobromobenzene, 2-fluoro-4-chlorobromobenzene, 2-bromo-5-chlorofluorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 1-bromo-2-fluoro-4-chlorobenzene, 2-bromo-5-chloro-1-fluorobenzene, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209, acmc-1bovk, 2-bromo-5-chlorofluorbenzene
137275
1-bromo-4-chloro-2-fluorobenzene
FC1=CC(Cl)=CC=C1Br

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5540 to 1.5560
Quantity 5 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 209.44
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-chloro-2-fluorobenzene, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'