missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(1,1-difluoroethyl)benzene, 95%, Thermo Scientific Chemicals

Catalog Number p-4228432
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1000994-95-5
C8H7BrF2
221.05
MFCD11110333
QXIBKCFAFRHORF-UHFFFAOYSA-N
1-bromo-4-1,1-difluoroethyl benzene, 1-bromo-4-1,1-difluoro-ethyl-benzene, benzene, 1-bromo-4-1,1-difluoroethyl, 4-1,1-difluoroethyl bromobenzene, bromo-4-1,1-difluoroethyl benzene, 1-bromo-4-1,1-difluoroethyl-benzene, 1-4-bromophenyl-1,1-difluoroethane
45158847
1-bromo-4-(1,1-difluoroethyl)benzene
CC(C1=CC=C(C=C1)Br)(F)F
This item is not returnable. View return policy
1000994-95-5
C8H7BrF2
221.05
MFCD11110333
QXIBKCFAFRHORF-UHFFFAOYSA-N
1-bromo-4-1,1-difluoroethyl benzene, 1-bromo-4-1,1-difluoro-ethyl-benzene, benzene, 1-bromo-4-1,1-difluoroethyl, 4-1,1-difluoroethyl bromobenzene, bromo-4-1,1-difluoroethyl benzene, 1-bromo-4-1,1-difluoroethyl-benzene, 1-4-bromophenyl-1,1-difluoroethane
45158847
1-bromo-4-(1,1-difluoroethyl)benzene
CC(C1=CC=C(C=C1)Br)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Brown
Boiling Point 80.0°C to 82.0°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 221.05
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 1-Bromo-4-(1, 1-difluoroethyl)benzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-4-(1,1-difluoroethyl)benzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'