missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals

Catalog Number p-3735687
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
109-70-6
C3H6BrCl
157.44
MFCD00000998
MFESCIUQSIBMSM-UHFFFAOYSA-N
trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane
8006
1-bromo-3-chloropropane
C(CCl)CBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
109-70-6
C3H6BrCl
157.44
MFCD00000998
MFESCIUQSIBMSM-UHFFFAOYSA-N
trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane
8006
1-bromo-3-chloropropane
C(CCl)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -59.0°C
Density 1.5900g/mL
Boiling Point 142.0°C to 145.0°C
Flash Point 81°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3Br
Refractive Index 1.4850 to 1.4870
Quantity 50 mL
Beilstein 01, 109
Specific Gravity 1.59
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,ether and chloroform
Formula Weight 157.44
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-chloropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-3-chloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'