missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-5-fluorobenzene, 98%, Acros Organics

Brand:  Acros Organics 448751000

Code : NEW

Additional Details : CAS Number : 33863-76-2 Weight : 0.10000kg


Quantity 100g

 View more versions of this product

Product Code. 13438618

  • £316.60 / 100g

Estimated Shipment date
from Supplier 15-06-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

33863-76-2
209.45
3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene
1-bromo-3-chloro-5-fluorobenzene
C6H3BrClF
GGMDFPMASIXEIR-UHFFFAOYSA-N
2736223
C1=C(C=C(C=C1Cl)Br)F

Specifications

1-Bromo-3-chloro-5-fluorobenzene
Glass bottle
72°C (5.0mmHg)
33863-76-2
C6H3BrClF
3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene
C1=C(C=C(C=C1Cl)Br)F
209.45
209.45
1.7200g/mL
1.72
100g
97.5% min. (GC)
BrC6H3(Cl)F
GGMDFPMASIXEIR-UHFFFAOYSA-N
1-bromo-3-chloro-5-fluorobenzene
2736223
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Bromo-3-chloro-5-fluorobenzene

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents