missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog Number 10510532
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19752-55-7
C6H3BrCl2
225.9
MFCD00000584
DZHFFMWJXJBBRG-UHFFFAOYSA-N
3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
29766
1-bromo-3,5-dichlorobenzene
C1=C(C=C(C=C1Cl)Br)Cl
This item is not returnable. View return policy
19752-55-7
C6H3BrCl2
225.9
MFCD00000584
DZHFFMWJXJBBRG-UHFFFAOYSA-N
3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
29766
1-bromo-3,5-dichlorobenzene
C1=C(C=C(C=C1Cl)Br)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 74°C to 77°C
Boiling Point 232°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 225.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3, 5-dichlorobenzene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'