missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,4-dichlorobenzene, 98+%, Thermo Scientific Chemicals
32.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Melting Point | 23.0°C to 24.0°C |
Color | Yellow |
Boiling Point | 140.0°C to 142.0°C (30.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Glass bottle |
Linear Formula | BrC6H3Cl2 |
Refractive Index | 1.5986 to 1.6006 |
Show More |
Safety and Handling
Product Identifier
- 1-Bromo-3,4-dichlorobenzene
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Skin sensitiser Category 1
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H335-May cause respiratory irritation.
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'