missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,3-dimethylbutan-2-one, Tech., Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Linear Formula | (CH3)3CCOCH2Br |
Quantity | 100 g |
Formula Weight | 179.06 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 1-Bromo-3, 3-dimethylbutan-2-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'