missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals

Catalog Number p-7027249
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
78-77-3
C4H9Br
137.02
MFCD00000217
HLVFKOKELQSXIQ-UHFFFAOYSA-N
isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
6555
1-bromo-2-methylpropane
CC(C)CBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
78-77-3
C4H9Br
137.02
MFCD00000217
HLVFKOKELQSXIQ-UHFFFAOYSA-N
isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
6555
1-bromo-2-methylpropane
CC(C)CBr

1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

Solubility
Slightly soluble in water. Miscible with alcohol, ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point -118°C
Density 1.255
Boiling Point 90°C to 92°C
Flash Point 18°C (64°F)
Refractive Index 1.435
Quantity 250 g
UN Number UN2342
Beilstein 1730915
Merck Index 14,5135
Solubility Information Slightly soluble in water; Miscible with alcohol,ether.
Formula Weight 137.02
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-methylpropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'