missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-iodobenzene, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number 10494985
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
583-55-1
C6H4BrI
282.9
MFCD00001030
OIRHKGBNGGSCGS-UHFFFAOYSA-N
2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
11415
1-bromo-2-iodobenzene
C1=CC=C(C(=C1)Br)I
583-55-1
C6H4BrI
282.9
MFCD00001030
OIRHKGBNGGSCGS-UHFFFAOYSA-N
2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
11415
1-bromo-2-iodobenzene
C1=CC=C(C(=C1)Br)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 9°C to 10°C
Color Yellow
Density 2.2030g/mL
Boiling Point 120°C to 121°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4I
Refractive Index 1.6620 to 1.664
Quantity 25 g
Beilstein 05, 223
Fieser 01,77
Specific Gravity 2.203
Stabilizer copper
Formula Weight 282.9
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2-iodobenzene, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'