missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-hexadecanone, 97%, Thermo Scientific Chemicals

Catalog Number p-7030307
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
21436-52-2
C16H31BrO
319.327
MFCD07777089
ICZYLTOWIBJLIK-UHFFFAOYSA-N
1-bromo-2-hexadecanone, acmc-1cipg, 2-hexadecanone,1-bromo
14843414
1-bromohexadecan-2-one
CCCCCCCCCCCCCCC(=O)CBr
This item is not returnable. View return policy
21436-52-2
C16H31BrO
319.327
MFCD07777089
ICZYLTOWIBJLIK-UHFFFAOYSA-N
1-bromo-2-hexadecanone, acmc-1cipg, 2-hexadecanone,1-bromo
14843414
1-bromohexadecan-2-one
CCCCCCCCCCCCCCC(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 54°C to 59°C
Quantity 10 g
Formula Weight 319.32
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-hexadecanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'