missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog Number 15438048
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
185331-69-5
C6H3BrFNO2
220.00
MFCD05865088
AZYQMHQOHNGPRR-UHFFFAOYSA-N
4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
2756994
1-bromo-2-fluoro-4-nitrobenzene
[O-][N+](=O)C1=CC(F)=C(Br)C=C1
This item is not returnable. View return policy
185331-69-5
C6H3BrFNO2
220.00
MFCD05865088
AZYQMHQOHNGPRR-UHFFFAOYSA-N
4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
2756994
1-bromo-2-fluoro-4-nitrobenzene
[O-][N+](=O)C1=CC(F)=C(Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 88°C to 94°C
Quantity 25 g
Formula Weight 220
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Bromo-2-fluoro-4-nitrobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'