missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylbutane, 97%, Thermo Scientific Chemicals

Catalog Number p-73870
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
3814-34-4
C6H13Br
165.08
MFCD00000219
KKGUMGWNFARLSL-UHFFFAOYSA-N
1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane
77432
3-(bromomethyl)pentane
CCC(CC)CBr
This item is not returnable. View return policy
3814-34-4
C6H13Br
165.08
MFCD00000219
KKGUMGWNFARLSL-UHFFFAOYSA-N
1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane
77432
3-(bromomethyl)pentane
CCC(CC)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 143°C to 144°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2CHCH2Br
Refractive Index 1.4510 to 1.453
Quantity 5 mL
Specific Gravity 1.17
Formula Weight 165.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethylbutane, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'