missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(difluoromethyl)benzene, 97%, Thermo Scientific™

Catalog Number 10812421
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
845866-82-2
C7H5BrF2
207.02
RMYPQIOCZSWSDG-UHFFFAOYSA-N
2761181
1-bromo-2-(difluoromethyl)benzene
C1=CC=C(C(=C1)C(F)F)Br
This item is not returnable. View return policy
845866-82-2
C7H5BrF2
207.02
RMYPQIOCZSWSDG-UHFFFAOYSA-N
2761181
1-bromo-2-(difluoromethyl)benzene
C1=CC=C(C(=C1)C(F)F)Br
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 74°C to 76°C (17.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Synonym 1-bromo-2-difluoromethyl benzene, 2-difluoromethyl bromobenzene, 2-bromophenyl difluoro methane, 1-bromo-2-difluoromethyl-benzene, benzene,1-bromo-2-difluoromethyl, 1-bromo-2-difluoromethylbenzene, benzene, 1-bromo-2-difluoromethyl
Formula Weight 207.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-(difluoromethyl)benzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2-(difluoromethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'