missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(difluoromethyl)benzene, 97%, ACROS Organics™

Brand:  Acros Organics 432540250

Code : 29

Additional Details : CAS Number : 845866-82-2 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 10462560

  • £451.00 / 25g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

1-Bromo-2-(difluoromethyl)benzene
96.0
≥96.0%
C7H5BrF2
845866-82-2
100.0
97%
207.02

Specifications

1-Bromo-2-(difluoromethyl)benzene
96.0
Authentic
≥96.0%
C7H5BrF2
Clear colorless
845866-82-2
100.0
Liquid
97%
207.02
25g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Bromo-2-(difluoromethyl)benzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS