missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,6-dichlorobenzene, 99%, Thermo Scientific™

Catalog Number p-73856
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19393-92-1
MFCD00000574
UWOIDOQAQPUVOH-UHFFFAOYSA-N
1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
29568
2-bromo-1,3-dichlorobenzene
C1=CC(=C(C(=C1)Cl)Br)Cl
This item is not returnable. View return policy
19393-92-1
MFCD00000574
UWOIDOQAQPUVOH-UHFFFAOYSA-N
1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
29568
2-bromo-1,3-dichlorobenzene
C1=CC(=C(C(=C1)Cl)Br)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 64.0°C to 67.0°C
Color White to Yellow
Boiling Point 242.0°C (765.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H3BrCl2
Linear Formula BrC6H3Cl2
Quantity 5 g
Beilstein 05, 210
Molecular Weight (g/mol) 225.9
Formula Weight 225.9
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Bromo-2, 6-dichlorobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2,6-dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'