missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dimethoxybenzene, 98%, Thermo Scientific™

Catalog Number p-73813
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
17715-69-4
C8H9BrO2
217.06
MFCD00009844
NIUZVSQOXJIHBL-UHFFFAOYSA-N
2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
87266
1-bromo-2,4-dimethoxybenzene
COC1=CC=C(Br)C(OC)=C1
This item is not returnable. View return policy
17715-69-4
C8H9BrO2
217.06
MFCD00009844
NIUZVSQOXJIHBL-UHFFFAOYSA-N
2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
87266
1-bromo-2,4-dimethoxybenzene
COC1=CC=C(Br)C(OC)=C1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 25°C to 26°C
Color Yellow
Density 1.5070g/mL
Boiling Point 153°C to 155°C (18.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OCH3)Br
Refractive Index 1.5718 to 1.5738
Quantity 5 mL
Beilstein 06, II, 819
Specific Gravity 1.507
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 217.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 4-dimethoxybenzene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'