missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7082049
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1193-72-2
C6H3BrCl2
225.894
MFCD00018333
ISHYFWKKWKXXPL-UHFFFAOYSA-N
2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l
70947
1-bromo-2,4-dichlorobenzene
C1=CC(=C(C=C1Cl)Cl)Br
This item is not returnable. View return policy
1193-72-2
C6H3BrCl2
225.894
MFCD00018333
ISHYFWKKWKXXPL-UHFFFAOYSA-N
2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l
70947
1-bromo-2,4-dichlorobenzene
C1=CC(=C(C=C1Cl)Cl)Br

1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 26°C to 27°C
Boiling Point 235°C
Quantity 5 g
Beilstein 1635733
Solubility Information Slightly soluble in water.
Formula Weight 225.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,4-dichlorobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'