missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5-trifluorobenzene, 98%, Thermo Scientific™

Catalog Number 11954551
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
133739-70-5
C6H2BrF3
210.98
MFCD00012232
XSMLLZPSNLQCQU-UHFFFAOYSA-N
2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
2736399
1-bromo-2,3,5-trifluorobenzene
FC1=CC(F)=C(F)C(Br)=C1
This item is not returnable. View return policy
133739-70-5
C6H2BrF3
210.98
MFCD00012232
XSMLLZPSNLQCQU-UHFFFAOYSA-N
2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
2736399
1-bromo-2,3,5-trifluorobenzene
FC1=CC(F)=C(F)C(Br)=C1
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 143°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 210.98
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 3, 5-trifluorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2,3,5-trifluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'