missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-pyrrolo[3,2-c]pyridine, 95%

Brand:  Alfa Aesar™ H64319.03

Code : NEW

Additional Details : CAS Number : 148760-75-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451469

  • £65.72 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-pyrrolo[3,2-c]pyridine
218.26
5866349
148760-75-2
MFCD11976180
tert-Butyl pyrrolo[3,2-c]pyridine-1-carboxylate

Specifications

1-Boc-pyrrolo[3,2-c]pyridine
Liquid
218.26
5866349
tert-Butyl pyrrolo[3,2-c]pyridine-1-carboxylate
148760-75-2
95%
MFCD11976180
Pale yellow
1g
Videos
SDS