Learn More
Alfa Aesar™ 1-Boc-pyrrolo[3,2-c]pyridine, 95%
Brand: Alfa Aesar™ H64319.06
Code : NEW
Additional Details : CAS Number : 148760-75-2 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
1-Boc-pyrrolo[3,2-c]pyridine | |
218.26 | |
5866349 |
148760-75-2 | |
MFCD11976180 | |
tert-Butyl pyrrolo[3,2-c]pyridine-1-carboxylate |
Specifications
1-Boc-pyrrolo[3,2-c]pyridine | |
Liquid | |
218.26 | |
5866349 | |
tert-Butyl pyrrolo[3,2-c]pyridine-1-carboxylate |
148760-75-2 | |
95% | |
MFCD11976180 | |
Pale yellow | |
5g |