missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-isonipecotamide, ≥97%

Brand:  Alfa Aesar™ H64657.06

Code : NEW

Additional Details : CAS Number : 91419-48-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15431949

  • £26.39 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-isonipecotamide
228.29
5533475
91419-48-6
MFCD02180953
1-Boc-piperidine-4-carboxamide; tert-Butyl 4-carbamoylpiperidine-1-carboxylate

Specifications

1-Boc-isonipecotamide
Solid
228.29
163° to 164°C
1-Boc-piperidine-4-carboxamide; tert-Butyl 4-carbamoylpiperidine-1-carboxylate
91419-48-6
≥97%
MFCD02180953
5533475
5g
Videos
SDS