missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-D-tryptophan, 95%

Brand:  Alfa Aesar™ H62809.03

Code : NEW

Additional Details : CAS Number : 146645-63-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469278

  • £20.99 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-D-tryptophan
304.34
H-Trp(Boc)-OH
146645-63-8
MFCD00237294

Specifications

1-Boc-D-tryptophan
Solid
304.34
H-Trp(Boc)-OH
146645-63-8
95%
MFCD00237294
1g
Videos
SDS