missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(Boc-amino)cyclobutanecarboxylic acid, 97%

Brand:  Alfa Aesar™ H62888.06

Code : NEW

Additional Details : CAS Number : 120728-10-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449338

  • £147.50 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(Boc-amino)cyclobutanecarboxylic acid
215.24
7994003
120728-10-1
MFCD02682623
1-tert-Butoxycarbonylaminocyclobutanecarboxylic acid; Boc-cyclovaline

Specifications

1-(Boc-amino)cyclobutanecarboxylic acid
Solid
129° to 133°C
MFCD02682623
7994003
120728-10-1
97%
215.24
5g
1-tert-Butoxycarbonylaminocyclobutanecarboxylic acid; Boc-cyclovaline
Videos
SDS