missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63394.03

Code : NEW

Additional Details : CAS Number : 166104-19-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15420069

  • £47.81 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-5-nitroindole
262.26
10344204
166104-19-4
MFCD09953094
tert-Butyl 5-nitroindole-1-carboxylate; 5-Nitroindole-1-carboxylic acid tert-butyl ester

Specifications

1-Boc-5-nitroindole
Solid
262.26
10344204
1g
166104-19-4
97%
MFCD09953094
tert-Butyl 5-nitroindole-1-carboxylate; 5-Nitroindole-1-carboxylic acid tert-butyl ester
Videos
SDS