missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-4-piperidone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
1-Boc-4-piperidone is used as a pharmaceutical intermediate. It is involved in the preparation of 1-piperidin-4-yl-substituted butyro- and valerolactams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-4-piperidone is used as a pharmaceutical intermediate. It is involved in the preparation of 1-piperidin-4-yl-substituted butyro- and valerolactams.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and metals.
1-Boc-4-piperidone is used as a pharmaceutical intermediate. It is involved in the preparation of 1-piperidin-4-yl-substituted butyro- and valerolactams.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and metals.
Specifications
Melting Point | 72°C to 76°C |
Quantity | 10 g |
Beilstein | 3650236 |
Solubility Information | Soluble in methanol. Slightly soluble in water. |
Formula Weight | 199.25 |
Percent Purity | 99% |
Chemical Name or Material | 1-Boc-4-piperidone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'