missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-cyano-4-(4-morpholinyl)piperidine, 97%

Brand:  Alfa Aesar™ H63252.03

Code : NEW

Additional Details : CAS Number : 1148003-98-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489848

  • £92.01 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-4-cyano-4-(4-morpholinyl)piperidine
295.38
tert-Butyl 4-cyano-4-(4-morpholinyl)piperidine-1-carboxylate; tert-Butyl 4-cyano-4-(4-morpholinyl)piperidine-1-carboxylate
1148003-98-8
MFCD12827546

Specifications

1-Boc-4-cyano-4-(4-morpholinyl)piperidine
Solid
295.38
1g
1148003-98-8
97%
MFCD12827546
tert-Butyl 4-cyano-4-(4-morpholinyl)piperidine-1-carboxylate; tert-Butyl 4-cyano-4-(4-morpholinyl)piperidine-1-carboxylate
Videos
SDS