missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-(4-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63615.03

Code : NEW

Additional Details : CAS Number : 182618-86-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400409

  • £40.60 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-4-(4-nitrophenyl)piperazine
307.34
tert-Butyl 4-(4-nitrophenyl)piperazine-1-carboxylate; 4-(4-Nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
182618-86-6
MFCD02187326

Specifications

1-Boc-4-(4-nitrophenyl)piperazine
Solid
307.34
tert-Butyl 4-(4-nitrophenyl)piperazine-1-carboxylate; 4-(4-Nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
182618-86-6
97%
MFCD02187326
1g
Videos
SDS