missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63438.03

Code : NEW

Additional Details : CAS Number : 874814-94-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410139

  • £50.70 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine
325.34
18325431
874814-94-5
MFCD06408840
tert-Butyl 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylate; 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester

Specifications

1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine
Solid
325.34
1g
tert-Butyl 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylate; 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
874814-94-5
97%
MFCD06408840
18325431
Videos
SDS