missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-(4-chloro-2-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63592.03

Code : NEW

Additional Details : CAS Number : 874814-97-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430369

  • £50.70 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-4-(4-chloro-2-nitrophenyl)piperazine
341.79
tert-Butyl 4-(4-chloro-2-nitrophenyl)-piperazine-1-carboxylate; 4-(4-Chloro-2-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester
874814-97-8
18477062

Specifications

1-Boc-4-(4-chloro-2-nitrophenyl)piperazine
Solid
341.79
18477062
874814-97-8
97%
1g
tert-Butyl 4-(4-chloro-2-nitrophenyl)-piperazine-1-carboxylate; 4-(4-Chloro-2-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester
Videos
SDS