missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-(3-hydroxypropyl)piperazine, 97%

Brand:  Alfa Aesar™ H55547.14

Code : NEW

Additional Details : CAS Number : 132710-90-8 Weight : 0.02500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15441848

  • £185.50 / 25g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-4-(3-hydroxypropyl)piperazine
132710-90-8
244.33
MFCD06798090

Specifications

1-Boc-4-(3-hydroxypropyl)piperazine
Solid
73° to 77°C
MFCD06798090
132710-90-8
97%
244.33
25g
Videos
SDS