Alfa Aesar™ 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H55598.14

25GR 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine, 97%

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 261925-94-4 Weight: 0.02500kg


Quantity 25g

Product Code. 15441918

Quantity Price
1 £ 297.01 / 25g
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine
CAS 261925-94-4
Formula Weight 334.42
MDL Number MFCD07369778
UN Number UN2811
Synonym 1-tert-Butoxycarbonyl-4-[3-(ethoxycarbonyl)phenyl]piperazine

Specification

Formula Weight 334.42
Melting Point 62° to 66°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine
Physical Form Powder
Assay 97%
MDL Number MFCD07369778
Quantity 25g
CAS 261925-94-4
UN Number UN2811
Color Yellow to orange
Synonym 1-tert-Butoxycarbonyl-4-[3-(ethoxycarbonyl)phenyl]piperazine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.