Learn More
Alfa Aesar™ 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine, 97%
Brand: Alfa Aesar™ H55598.06
Code : NEW
Additional Details : CAS Number : 261925-94-4 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine | |
334.42 | |
UN2811 |
261925-94-4 | |
MFCD07369778 | |
1-tert-Butoxycarbonyl-4-[3-(ethoxycarbonyl)phenyl]piperazine |
Specifications
1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine | |
Powder | |
62° to 66°C | |
MFCD07369778 | |
UN2811 | |
1-tert-Butoxycarbonyl-4-[3-(ethoxycarbonyl)phenyl]piperazine |
261925-94-4 | |
97% | |
334.42 | |
5g | |
Yellow to orange |