missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-(2-aminophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63364.06

Code : NEW

Additional Details : CAS Number : 170017-74-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15400029

  • £208.82 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-Boc-4-(2-aminophenyl)piperazine
277.36
tert-Butyl 4-(2-aminophenyl)piperazine-1-carboxylate; 4-(2-Aminophenyl)piperazine-1-carboxylic acid tert-butyl ester
170017-74-0
MFCD04115046

Specifications

1-Boc-4-(2-aminophenyl)piperazine
Solid
120° to 122°C
MFCD04115046
tert-Butyl 4-(2-aminophenyl)piperazine-1-carboxylate; 4-(2-Aminophenyl)piperazine-1-carboxylic acid tert-butyl ester
170017-74-0
97%
277.36
5g
Videos
SDS