missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-piperidone, 97%, Thermo Scientific Chemicals

Catalog Number p-7047463
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
98977-36-7
C10H17NO3
199.25
MFCD01631193
RIFXIGDBUBXKEI-UHFFFAOYSA-N
1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
2756825
tert-butyl 3-oxopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(=O)C1
This item is not returnable. View return policy
98977-36-7
C10H17NO3
199.25
MFCD01631193
RIFXIGDBUBXKEI-UHFFFAOYSA-N
1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
2756825
tert-butyl 3-oxopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(=O)C1

1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Melting Point 37°C to 39°C
Quantity 1 g
Beilstein 5936353
Solubility Information It is insoluble in water.
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material 1-Boc-3-piperidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'