missing translation for 'onlineSavingsMsg'
Learn More

1-[Bis(4-fluorophenyl)methyl]piperazine, 97%, Alfa Aesar™

Brand:  Alfa Aesar H55173.06

Code : NEW

Additional Details : CAS Number : 27469-60-9 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15401338

  • £56.96 / 5g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

27469-60-9
288.342
TTXIFFYPVGWLSE-UHFFFAOYSA-N
152932
C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
C17H18F2N2
MFCD00038660
1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
1-[bis(4-fluorophenyl)methyl]piperazine

Specifications

1-[Bis(4-fluorophenyl)methyl]piperazine
5g
C17H18F2N2
UN2811
1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
288.342
288.34
88°C to 92°C
27469-60-9
MFCD00038660
620754
TTXIFFYPVGWLSE-UHFFFAOYSA-N
1-[bis(4-fluorophenyl)methyl]piperazine
152932
97%
Videos
SDS Documents
Certificates