missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxycarbonyl-4-piperidone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7026889
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
19099-93-5
C13H15NO3
233.267
MFCD00673144
VZOVOHRDLOYBJX-UHFFFAOYSA-N
1-cbz-4-piperidone, n-benzyloxycarbonyl-4-piperidone, 1-cbz-piperidin-4-one, n-cbz-4-piperidone, benzyl 4-oxo-1-piperidinecarboxylate, 1-benzyloxycarbonyl-4-piperidinone, 1-z-4-piperidone, 1-piperidinecarboxylic acid, 4-oxo-, phenylmethyl ester, 1-carbobenzoxy-4-piperidone, 1-benzyloxycarbonyl-4-piperidone
643496
benzyl 4-oxopiperidine-1-carboxylate
C1CN(CCC1=O)C(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
19099-93-5
C13H15NO3
233.267
MFCD00673144
VZOVOHRDLOYBJX-UHFFFAOYSA-N
1-cbz-4-piperidone, n-benzyloxycarbonyl-4-piperidone, 1-cbz-piperidin-4-one, n-cbz-4-piperidone, benzyl 4-oxo-1-piperidinecarboxylate, 1-benzyloxycarbonyl-4-piperidinone, 1-z-4-piperidone, 1-piperidinecarboxylic acid, 4-oxo-, phenylmethyl ester, 1-carbobenzoxy-4-piperidone, 1-benzyloxycarbonyl-4-piperidone
643496
benzyl 4-oxopiperidine-1-carboxylate
C1CN(CCC1=O)C(=O)OCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38°C to 41°C
Density 1.172
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 233.27
Percent Purity ≥98%
Chemical Name or Material 1-Benzyloxycarbonyl-4-piperidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'