missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxycarbonyl-3-piperidone, 97+%, Thermo Scientific Chemicals

Catalog Number p-7055801
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
61995-20-8
C13H15NO3
233.27
MFCD03001723
ALXLNFWWLXCXSK-UHFFFAOYSA-N
1-n-cbz-3-piperidone, 1-cbz-3-piperidinone, 1-cbz-3-piperidone, n-cbz-3-piperidone, n-benzyloxycarbonyl-3-piperidone, 1-piperidinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxo-piperidine-1-carboxylic acid benzyl ester, 1-benzyloxycarbonyl-3-piperidone, 3-oxopiperidine-1-carboxylic acid benzyl ester, pubchem24305
1514169
benzyl 3-oxopiperidine-1-carboxylate
O=C(OCC1=CC=CC=C1)N1CCCC(=O)C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
61995-20-8
C13H15NO3
233.27
MFCD03001723
ALXLNFWWLXCXSK-UHFFFAOYSA-N
1-n-cbz-3-piperidone, 1-cbz-3-piperidinone, 1-cbz-3-piperidone, n-cbz-3-piperidone, n-benzyloxycarbonyl-3-piperidone, 1-piperidinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxo-piperidine-1-carboxylic acid benzyl ester, 1-benzyloxycarbonyl-3-piperidone, 3-oxopiperidine-1-carboxylic acid benzyl ester, pubchem24305
1514169
benzyl 3-oxopiperidine-1-carboxylate
O=C(OCC1=CC=CC=C1)N1CCCC(=O)C1

1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 1464624
Solubility Information Slightly soluble in water.
Formula Weight 233.27
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1-Benzyloxycarbonyl-3-piperidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'