missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyloxycarbonyl-3-piperidone, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Quantity | 5 g |
Beilstein | 1464624 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 233.27 |
Percent Purity | ≥97% |
Physical Form | Liquid |
Chemical Name or Material | 1-Benzyloxycarbonyl-3-piperidone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'