missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylimidazole, 99%, ACROS Organics™

Brand:  Acros Organics 152251000

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 4238-71-5 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10549433

  • £183.40 / 100g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

1-Benzylimidazole
98.5
99%
C10H10N2
MFCD00005296
4238-71-5
100.0
99%
158.2

Specifications

1-Benzylimidazole
98.5
Crystals
99%
158.2
23, 48
Glass bottle
Brownish
100g
4238-71-5
100.0
99%
C10H10N2
MFCD00005296
Authentic
310.0°C
68.0° to 71.0°C
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Benzylimidazole

Documentation

Technical Documents