missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylimidazole, 99%, Thermo Scientific Chemicals

Catalog Number p-3777397
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4238-71-5
C10H10N2
158.20
MFCD00005296
KKKDZZRICRFGSD-UHFFFAOYSA-N
1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole
77918
1-benzylimidazole
C(N1C=CN=C1)C1=CC=CC=C1
This item is not returnable. View return policy
4238-71-5
C10H10N2
158.20
MFCD00005296
KKKDZZRICRFGSD-UHFFFAOYSA-N
1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole
77918
1-benzylimidazole
C(N1C=CN=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 68.0°C to 71.0°C
Color Brown
Boiling Point 310.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 48
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol and acetone
Formula Weight 158.2
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material 1-Benzylimidazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'